Dibutyl Itaconate ≥97.0% (by GC) stabilised
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Itaconic Acid Dibutyl Ester, Dibutyl 2-methylenesuccinate
Stabilisatie: Stabilised with hydroquinone (HQ)
I0201-25G I0201-500G
TCIAI0201-25GEA 23.1 EUR
TCIAI0201-25G TCIAI0201-500G
Dibutyl Itaconate ≥97.0% (by GC) stabilised
Dibutyl Itaconate
Formule: C₁₃H₂₂O₄
MW: 242.32 g/mol
Kookpunt: 190°C
Vlampunt: 138°C
MDL Nummer: MFCD00027211
CAS nummer: 2155-60-4
EINECS: 218-451-9
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %