Sodium isoascorbate monohydrate 98,0 Titrimetric

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: Isoascorbic acid Sodium salt, Sodium Erythorbate, Sodium (R)-2-((R)-1,2-dihydroxyethyl)-4-hydroxy-5-oxo-2,5-dihydrofuran-3-olate, Erythorbic acid Sodium salt, Sodium D-Araboascorbate, D-Araboascorbic acid Sodium salt

I0232-25G I0232-500G
TCIAI0232-25GEA 17.6 EUR
TCIAI0232-25G TCIAI0232-500G
Sodium isoascorbate monohydrate 98,0 Titrimetric
Sodium isoascorbate monohydrate
Formule: C₆H₈O₆
MW: 198.11 g/mol
MDL Nummer: MFCD00067298
CAS nummer: 6381-77-7
EINECS: 228-973-9

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (Iodometric Titration) min. 98.0 %
Specific rotation [a]20/D +96.0 to +98.0 deg (C=10, H2O)
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR