Isobutyraldehyde 2,4-Dinitrophenylhydrazone ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

I0438-1G I0438-10G
TCIAI0438-1GEA 23.1 EUR
TCIAI0438-1G TCIAI0438-10G
Isobutyraldehyde 2,4-Dinitrophenylhydrazone ≥98.0% (by titrimetric analysis)
Isobutyraldehyde 2,4-Dinitrophenylhydrazone
Formule: C₁₀H₁₂N₄O₄
MW: 252.23 g/mol
MDL Nummer: MFCD00191659
CAS nummer: 2057-82-1
UN: 1325
ADR: 4.1 III

Order Now

Specification Test Results

Appearance Light yellow to Brown powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 183.0 to 188.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR