Methyl (S)-(-)-2-isocyanatopropionate ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
I0465-1G
I0465-5G
TCIAI0465-1GEA
279
EUR
TCIAI0465-1G
TCIAI0465-5G
Methyl (S)-(-)-2-isocyanatopropionate ≥98.0% (by GC)
Methyl (S)-(-)-2-isocyanatopropionate
Formule:
C₅H₇NO₃ Kookpunt: 61°C |
MDL Nummer:
MFCD00191530 CAS nummer: 30293-82-4 UN: 2206 ADR: 6.1,III |
Specification Test Results
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 98.0 % |
Specific rotation [a]20/D | -20.0 to -25.0 deg (neat) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...