2-Indancarboxylic acid ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

I0532-1G I0532-5G
TCIAI0532-1GEA 27.5 EUR
TCIAI0532-1G TCIAI0532-5G
2-Indancarboxylic acid ≥98.0% (by titrimetric analysis)
2-Indancarboxylic acid
Formule: C₁₀H₁₀O₂
Kookpunt: 192°C
MDL Nummer: MFCD00085095
CAS nummer: 25177-85-9

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 126.0 to 131.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR