N2-Isobutyrylguanosine, monohydrate

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: N-Isobutyrylguanosine, N-(9-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-6-oxo-6,9-dihydro-1H-purin-2-yl)isobutyramide

I0699-1G I0699-100MG
TCIAI0699-1GEA 460 EUR
TCIAI0699-1G TCIAI0699-100MG
N2-Isobutyrylguanosine, monohydrate
N2-Isobutyrylguanosine
Formule: C₁₄H₁₉N₅O₆
Smeltpunt: 148°C
MDL Nummer: MFCD00274102
CAS nummer: 64350-24-9

Order Now

Specification Test Results

Appearance White to almost white powder to crystal
Water 4.3 to 5.3 %
Elemental analysis (Nitrogen) 18.0 to 20.0% (calcd.on anh.substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR