N'-Isopropylidene-2-nitrobenzenesulphonohydrazide ≥98.0% (by HPLC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
I0777-5G
I0777-1G
TCIAI0777-5GEA
152
EUR
TCIAI0777-5G
TCIAI0777-1G
N'-Isopropylidene-2-nitrobenzenesulphonohydrazide ≥98.0% (by HPLC, titration analysis)
N'-Isopropylidene-2-nitrobenzenesulphonohydrazide
Formule:
C₉H₁₁N₃O₄S Smeltpunt: 136°C Bewaar temperatuur: Vriezer |
MDL Nummer:
MFCD09800525 CAS nummer: 6655-27-2 |
Specification Test Results
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...