2-Iodobenzyl alcohol ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

I0858-5G I0858-25G
TCIAI0858-5GEA 36.3 EUR
TCIAI0858-5G TCIAI0858-25G
2-Iodobenzyl alcohol ≥98.0% (by GC)
2-Iodobenzyl alcohol
Formule: C₇H₇IO
Kookpunt: 145°C
MDL Nummer: MFCD00004610
CAS nummer: 5159-41-1
EINECS: 225-933-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b866abe7e58fd6896b71b8aaa2de28f2355ea6f7&vid=b866abe7e58fd6896b71b8aaa2de28f2355ea6f7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 90.0 to 94.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR