2-Iodo-4-nitroaniline ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

I0878-1G I0878-5G
TCIAI0878-1GEA 45.1 EUR
TCIAI0878-1G TCIAI0878-5G
2-Iodo-4-nitroaniline ≥98.0% (by GC)
2-Iodo-4-nitroaniline
Formule: C₆H₅IN₂O₂
MW: 264.02 g/mol
MDL Nummer: MFCD00955758
CAS nummer: 6293-83-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4077286540b64737ef4d8a849aa35520a4d082b8&vid=4077286540b64737ef4d8a849aa35520a4d082b8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (GC) min. 98.0 %
Melting point 105.0 to 109.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR