Isopentyl hexanoate ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

I0899-25ML I0899-100ML
TCIAI0899-25MLEA 20.9 EUR
TCIAI0899-25ML TCIAI0899-100ML
Isopentyl hexanoate ≥98.0% (by GC)
Isopentyl hexanoate
Formule: C₁₁H₂₂O₂
Kookpunt: 98°C
Vlampunt: 88°C
MDL Nummer: MFCD00027280
CAS nummer: 2198-61-0
EINECS: 218-600-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd985a9e054af7aea1915cdadaf7abcaad19e6df&vid=fd985a9e054af7aea1915cdadaf7abcaad19e6df&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR