3-Iodo-2-methoxypyridine ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

I0924-1G I0924-5G
TCIAI0924-1GEA 40.7 EUR
TCIAI0924-1G TCIAI0924-5G
3-Iodo-2-methoxypyridine ≥98.0% (by GC, titration analysis)
3-Iodo-2-methoxypyridine
Formule: C₆H₆INO
Kookpunt: 85°C
Vlampunt: 110°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD03094946
CAS nummer: 112197-15-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e9c11544a3b210b377b5244e64a936f76a916585&vid=e9c11544a3b210b377b5244e64a936f76a916585&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Light yellow to Brown clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR