4-Amino-3-iodo-1H-pyrazolo[3,4-d]pyrimidine ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

I0941-1G I0941-5G
TCIAI0941-1GEA 64.9 EUR
TCIAI0941-1G TCIAI0941-5G
4-Amino-3-iodo-1H-pyrazolo[3,4-d]pyrimidine ≥98.0% (by HPLC, titration analysis)
4-Amino-3-iodo-1H-pyrazolo[3,4-d]pyrimidine
Formule: C₅H₄N₅I
MDL Nummer: MFCD03787931
CAS nummer: 151266-23-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2329c4b7b1fe460150efaeeaf8def193becf5e90&vid=2329c4b7b1fe460150efaeeaf8def193becf5e90&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR