Iguratimod ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

I0945-25MG I0945-250MG
TCIAI0945-25MGEA 18.7 EUR
TCIAI0945-25MG TCIAI0945-250MG
Iguratimod ≥98.0% (by HPLC, titration analysis)
Iguratimod
Formule: C₁₇H₁₄N₂O₆S
MDL Nummer: MFCD00882374
CAS nummer: 123663-49-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5172f6da54b8280c6056f3631b1ff52e28abe5f6&vid=5172f6da54b8280c6056f3631b1ff52e28abe5f6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 238.0 to 242.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR