Ibutilide hemifumarate ≥98.0% (by HPLC, total nitrogen)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
N-(4-(4-(Ethyl(heptyl)amino)-1-hydroxybutyl)phenyl)methanesulphonamide hemifumarate, Ibutilide fumarate
I0946-20MG
I0946-100MG
TCIAI0946-20MGEA
63.8
EUR
TCIAI0946-20MG
TCIAI0946-100MG
Ibutilide hemifumarate ≥98.0% (by HPLC, total nitrogen)
Ibutilide hemifumarate
Formule:
C₂₀H₃₆N₂O₃S MW: 442.62 g/mol |
MDL Nummer:
MFCD01715410 CAS nummer: 122647-32-9 Merck Index: 14,04883 |
|
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...