Isoxadifen-ethyl ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

I0956-5G I0956-25G
TCIAI0956-5GEA 78.1 EUR
TCIAI0956-5G TCIAI0956-25G
Isoxadifen-ethyl ≥98.0% (by GC)
Isoxadifen-ethyl
Formule: C₁₈H₁₇NO₃
MDL Nummer: MFCD03792846
CAS nummer: 163520-33-0
EINECS: 443-870-0
UN: 3077
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9c3e23f8f0c0aa9b0f3afb3a52b56ae3935136f4&vid=9c3e23f8f0c0aa9b0f3afb3a52b56ae3935136f4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 85.0 to 89.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR