5-Iodo-2-methoxybenzonitrile ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
I0960-1G
I0960-5G
TCIAI0960-1GEA
46.2
EUR
TCIAI0960-1G
TCIAI0960-5G
5-Iodo-2-methoxybenzonitrile ≥98.0% (by GC)
5-Iodo-2-methoxybenzonitrile
Formule:
C₈H₆INO |
MDL Nummer:
MFCD02683860 CAS nummer: 933672-32-3 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 122.0 to 126.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...