2-Iododibenzofuran ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

I0968-1G I0968-5G
TCIAI0968-1GEA 165 EUR
TCIAI0968-1G TCIAI0968-5G
2-Iododibenzofuran ≥98.0% (by GC)
2-Iododibenzofuran
Formule: C₁₂H₇IO
MDL Nummer: MFCD00092649
CAS nummer: 5408-56-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4a76789f1bc64f59e7717bb42b8d690b662e2bd&vid=c4a76789f1bc64f59e7717bb42b8d690b662e2bd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 110.0 to 114.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR