PluriSIn 1 ≥98.0% (by HPLC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

I0975-20MG I0975-100MG
TCIAI0975-20MGEA 63.8 EUR
TCIAI0975-20MG TCIAI0975-100MG
PluriSIn 1 ≥98.0% (by HPLC)
PluriSIn 1
Formule: C₁₂H₁₁N₃O
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD02344904
CAS nummer: 91396-88-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=256a2ab65b801358b89af7405f79242e1072d4f3&vid=256a2ab65b801358b89af7405f79242e1072d4f3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 18.70 to 20.70 %
Melting point 186.0 to 190.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR