2-(4-Iodophenyl)-1-phenylbenzimidazole ≥98.0% (by HPLC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
2-(4-Iodophenyl)-1-phenyl-1H-benzo[d]imidazole
I0983-200MG
I0983-1G
TCIAI0983-200MGEA
68.2
EUR
TCIAI0983-200MG
TCIAI0983-1G
2-(4-Iodophenyl)-1-phenylbenzimidazole ≥98.0% (by HPLC, titration analysis)
2-(4-Iodophenyl)-1-phenylbenzimidazole
Formule:
C₁₉H₁₃IN₂ |
MDL Nummer:
MFCD29089366 CAS nummer: 760212-42-8 |
|
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 178.0 to 182.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...