3-Iodophenyl isothiocyanate ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
3-Iodophenylisothiocyanate, 1-Iodo-3-isothiocyanatobenzene
I0985-1G
I0985-5G
TCIAI0985-1GEA
29.7
EUR
TCIAI0985-1G
TCIAI0985-5G
3-Iodophenyl isothiocyanate ≥98.0% (by GC)
3-Iodophenyl isothiocyanate
Formule:
C₇H₄INS MW: 261.09 g/mol Kookpunt: 296 Smeltpunt: 46 |
MDL Nummer:
MFCD00041074 CAS nummer: 3125-73-3 UN: 2923 ADR: 8 III |
|
Specification Test Results
Appearance | White to Orange to Green powder to lump |
Purity (GC) | min. 98.0 % |
Melting point | 45.0 to 49.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...