2-Isopropyl-5-methylanisole ≥96.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

I0996-5G I0996-25G
TCIAI0996-5GEA 40.7 EUR
TCIAI0996-5G TCIAI0996-25G
2-Isopropyl-5-methylanisole ≥96.0% (by GC)
2-Isopropyl-5-methylanisole
Formule: C₁₁H₁₆O
Kookpunt: 216°C
Vlampunt: 51°C
MDL Nummer: MFCD01674973
CAS nummer: 1076-56-8
EINECS: 214-063-9
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e625598d48d75d827fd5e3622e4c9df685e487a8&vid=e625598d48d75d827fd5e3622e4c9df685e487a8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 96.0 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR