Perfluoro-15-crown-5-ether ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
I1043-1G I1043-5G
TCIAI1043-1GEA 57.2 EUR
TCIAI1043-1G TCIAI1043-5G
Perfluoro-15-crown-5-ether ≥98.0% (by GC)
Perfluoro-15-crown-5-ether
Formule: C₁₀F₂₀O₅
Kookpunt: 146°C
Smeltpunt: -12°C
MDL Nummer: MFCD01320747
CAS nummer: 97571-69-2
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %