Sisomicin sulphate Salt ≥98.0% (by HPLC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
I1049-1G
I1049-250MG
TCIAI1049-1GEA
126
EUR
TCIAI1049-1G
TCIAI1049-250MG
Sisomicin sulphate Salt ≥98.0% (by HPLC)
Sisomicin sulphate Salt
Formule:
(C₁₉H₃₇N₅O₇)₂·5H₂SO₄ Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00058308 CAS nummer: 53179-09-2 EINECS: 258-414-4 Merck Index: 14,08550 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Specific rotation [a]20/D | +98.0 to +104.0 deg (C=1, H2O) |
Drying loss | max. 5.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...