Isobutyl methanesulphonate ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Methanesulphonic acid Isobutyl ester
I1062-1G I1062-5G
TCIAI1062-1GEA 36.3 EUR
TCIAI1062-1G TCIAI1062-5G
Isobutyl methanesulphonate ≥98.0% (by GC)
Isobutyl methanesulphonate
Formule: C₅H₁₂O₃S
MW: 152.21 g/mol
Kookpunt: 53°C
MDL Nummer: MFCD07363982
CAS nummer: 16156-53-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e51ed1168cae7bc497fafa1c6824874be689d1ab&vid=e51ed1168cae7bc497fafa1c6824874be689d1ab&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %