Isononyl acrylate (mixture of branched chain isomers) stabilised

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing


Stabilisatie: Stabilised with 4-methoxyphenol (MEHQ)

I1071-100ML I1071-500ML
TCIAI1071-100MLEA 48.4 EUR
TCIAI1071-100ML TCIAI1071-500ML
Isononyl acrylate (mixture of branched chain isomers) stabilised
Isononyl acrylate (mixture of branched chain isomers)
Formule: C₁₂H₂₂O₂
CAS nummer: 51952-49-9
EINECS: 257-544-9

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Average alkylchain length 8.5 to 9.4


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR