Isopropoxybenzene ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
I1073-5G
I1073-25G
TCIAI1073-5GEA
35.2
EUR
TCIAI1073-5G
TCIAI1073-25G
Isopropoxybenzene ≥98.0% (by GC)
Isopropoxybenzene
Formule:
C₉H₁₂O Kookpunt: 176°C Smeltpunt: –33°C |
MDL Nummer:
MFCD00048288 CAS nummer: 2741-16-4 EINECS: 220-370-9 |
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...