1-Isopropoxy-2-propanol ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
I1084-1G
I1084-5G
TCIAI1084-1GEA
116
EUR
TCIAI1084-1G
TCIAI1084-5G
1-Isopropoxy-2-propanol ≥98.0% (by GC)
1-Isopropoxy-2-propanol
Formule:
C₆H₁₄O₂ Kookpunt: 137°C |
MDL Nummer:
MFCD06252288 CAS nummer: 3944-36-3 EINECS: 223-533-2 UN: 1987 ADR: 3,III |
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...