Isopentylamine hydroiodide ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

I1095-1G I1095-5G
TCIAI1095-1GEA 28.6 EUR
TCIAI1095-1G TCIAI1095-5G
Isopentylamine hydroiodide ≥98.0% (by titrimetric analysis)
Isopentylamine hydroiodide
Formule: C₅H₁₃N·HI
MDL Nummer: MFCD33021473
CAS nummer: 2733412-76-3

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR