IWP-2 ≥96.0% (by HPLC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

I1097-10MG I1097-50MG
TCIAI1097-10MGEA 131 EUR
TCIAI1097-10MG TCIAI1097-50MG
IWP-2 ≥96.0% (by HPLC)
IWP-2
Formule: C₂₂H₁₈N₄O₂S₃
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD12912418
CAS nummer: 686770-61-6

Order Now

Specification Test Results

Appearance White to Yellow to Orange powder to crystal
Purity (HPLC) min. 96.0 area%
Elemental analysis (Nitrogen) 11.40 to 12.60 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR