ID-8 ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: 1-(4-Methoxyphenyl)-2-methyl-3-nitro-1H-indol-6-ol

I1108-10MG I1108-50MG
TCIAI1108-10MGEA 125 EUR
TCIAI1108-10MG TCIAI1108-50MG
ID-8 ≥98.0% (by GC)
ID-8
Formule: C₁₆H₁₄N₂O₄
Smeltpunt: 266°C
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD00593367
CAS nummer: 147591-46-6

Order Now

Specification Test Results

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR