Janus green B

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

J0002-25G J0002-5G
TCIAJ0002-25GEA 120 EUR
TCIAJ0002-25G TCIAJ0002-5G
Janus green B
Janus green B
Formule: C₃₀H₃₁ClN₆
MDL Nummer: MFCD00011758
CAS nummer: 2869-83-2
EINECS: 220-695-6
Merck Index: 14,05255

Order Now

Specification Test Results

Appearance Dark green to Dark blue to Black powder to crystal
Lambda max. 651.0 to 666.0 nm (EtOH (50%))
Absorbance (E1%1cm) min. 600 (EtOH (50%), 651.0 to 666.0 nm)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR