JK-P3 ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

J0012-25MG J0012-5MG
TCIAJ0012-25MGEA 361 EUR
TCIAJ0012-25MG TCIAJ0012-5MG
JK-P3 ≥98.0% (by HPLC, titration analysis)
JK-P3
Formule: C₁₈H₁₇N₃O₃
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD22666405
CAS nummer: 942655-44-9

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR