KN-93 ≥98.0% (by HPLC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
N-[2-[[[3-(4-Chlorophenyl)allyl](methyl)amino]methyl]phenyl]-N-(2-hydroxyethyl)-4-methoxybenzenesulphonamide, N-(2-(((3-(4-Chlorophenyl)allyl)(methyl)amino)methyl)phenyl)-N-(2-hydroxyethyl)-4-methoxybenzenesulphonamide
K0055-25MG
K0055-5MG
TCIAK0055-25MGEA
626
EUR
TCIAK0055-25MG
TCIAK0055-5MG
KN-93 ≥98.0% (by HPLC)
KN-93
Formule:
C₂₆H₂₉ClN₂O₄S MW: 501.04 g/mol Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00236424 CAS nummer: 139298-40-1 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Elemental analysis (Nitrogen) | 5.00 to 6.00 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...