Kaempferide ≥96.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

K0057-10MG K0057-50MG
TCIAK0057-10MGEA 67.1 EUR
TCIAK0057-10MG TCIAK0057-50MG
Kaempferide ≥96.0% (by GC)
Kaempferide
Formule: C₁₆H₁₂O₆
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00016771
CAS nummer: 491-54-3
EINECS: 207-738-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6f8f73adf6b9c79c63c0583a5bfb4c6282afc9c1&vid=6f8f73adf6b9c79c63c0583a5bfb4c6282afc9c1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 96.0 %
Melting point 227.0 to 231.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR