Kirenol ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

K0059-100MG K0059-25MG
TCIAK0059-100MGEA 370 EUR
TCIAK0059-100MG TCIAK0059-25MG
Kirenol ≥98.0% (by GC)
Kirenol
Formule: C₂₀H₃₄O₄
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00210519
CAS nummer: 52659-56-0

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 197.0 to 201.0 deg-C
Specific rotation (value) -32.0 to -29.0 deg


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR