N-Lauryldiethanolamine ≥95.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

L0020-25G L0020-5G
TCIAL0020-25GEA 174 EUR
TCIAL0020-25G TCIAL0020-5G
N-Lauryldiethanolamine ≥95.0% (by GC)
N-Lauryldiethanolamine
Formule: C₁₆H₃₅NO₂
Kookpunt: 195°C
MDL Nummer: MFCD00059002
CAS nummer: 1541-67-9
EINECS: 216-277-8

Order Now

Specification Test Results

Appearance White or Colorless to Light yellow to Light orange powder to lump to clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR