Lignin (dealkaline)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

L0045-25G L0045-500G
TCIAL0045-25GEA 22 EUR
TCIAL0045-25G TCIAL0045-500G
Lignin (dealkaline)
Lignin (dealkaline)
MDL Nummer: MFCD00147441
CAS nummer: 9005-53-2
EINECS: 232-682-2
Merck Index: 14,05487

Order Now

Specification Test Results

Appearance Dark green to Dark red to Black powder to crystal
Methoxyl Group 10.0 to 12.5 % (calcd.on anh.substance)
Water max. 20.0 %
Ignition residue (Sulfate) 10.0 to 20.0 % (calcd.on anh.substance)
pH 3.0 to 4.0 (50g/L, 25deg-C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR