Lacmoid

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

L0079-5G L0079-1G
TCIAL0079-5GEA 41.8 EUR
TCIAL0079-5G TCIAL0079-1G
Lacmoid
Lacmoid
MDL Nummer: MFCD00152579
CAS nummer: 42249-61-6
EINECS: 255-736-7
Merck Index: 14,05332

Order Now

Specification Test Results

Appearance Black powder to crystal
Lambda max. 591.0 to 595.0nm (pH 7.0 buffer sol.)
Absorbance (E1%1cm) min. 100 (pH 7.0 buffer sol., 591.0 to 595.0 nm)
Transition interval (pH) (red)4.4 to 6.2 (lavender)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR