Lignin (alkaline)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
L0082-500G
L0082-25G
TCIAL0082-500GEA
35.2
EUR
TCIAL0082-500G
TCIAL0082-25G
Lignin (alkaline)
Lignin (alkaline)
MDL Nummer:
MFCD00135572 CAS nummer: 8061-51-6 EINECS: 617-145-6 Merck Index: 14,05487 |
Specification Test Results
Appearance | Black powder to crystal |
pH | 8.0 to 10.0 (50g/L, 20 deg-C) |
Methoxyl Group | 10.0 to 12.0 % (calcd.on anh.substance) |
Ignition residue (Sulfate) | 20.0 to 29.0 % (calcd.on anh.substance) |
Water | max. 10.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...