Methyl lignocerate ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
L0112-1G
L0112-5G
TCIAL0112-1GEA
69.3
EUR
TCIAL0112-1G
TCIAL0112-5G
Methyl lignocerate ≥98.0% (by GC)
Methyl lignocerate
Formule:
C₂₅H₅₀O₂ Kookpunt: 232°C |
MDL Nummer:
MFCD00009351 CAS nummer: 2442-49-1 EINECS: 219-475-2 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 58.0 to 61.0 deg-C |
Solubility in hot EtOH | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...