Levoglucosenone ≥96.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
L0150-200MG
L0150-1G
TCIAL0150-200MGEA
159
EUR
TCIAL0150-200MG
TCIAL0150-1G
Levoglucosenone ≥96.0% (by GC)
Levoglucosenone
Formule:
C₆H₆O₃ Kookpunt: 113°C |
MDL Nummer:
MFCD00191664 CAS nummer: 37112-31-5 Merck Index: 14,05465 |
Specification Test Results
Appearance | White to Yellow to Orange clear liquid |
Purity (GC) | min. 96.0 % |
Specific rotation [a]20/D | -530 to -570 deg (C=1, CHCl3) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...