Lead(II) bromide for perovskite precursor
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Lead dibromide, Lead(II) bromide
L0288-5G L0288-25G L0288-1G
TCIAL0288-5GEA 74.8 EUR
TCIAL0288-5G TCIAL0288-25G TCIAL0288-1G
Lead(II) bromide for perovskite precursor
Lead(II) bromide
Formule: Br₂Pb
MW: 367.01 g/mol
Kookpunt: 916
Smeltpunt: 373°C
MDL Nummer: MFCD00011156
CAS nummer: 10031-22-8
EINECS: 233-084-4
UN: 2291
ADR: 6.1 III
Merck Index: 14,05403
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0df59e07414b9a39a79fae159c6201a8e5d8fd7a&vid=0df59e07414b9a39a79fae159c6201a8e5d8fd7a&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White to Almost white powder to crystaline