Bis(2-ethylhexyl)maleate ≥93.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: Maleic Acid Di(2-ethylhexyl) Ester, Maleic Acid Dioctyl Ester, Bis(2-ethylhexyl) maleate, Di(2-ethylhexyl) Maleate

M0011-25ML M0011-500ML
TCIAM0011-25MLEA 18.7 EUR
TCIAM0011-25ML TCIAM0011-500ML
Bis(2-ethylhexyl)maleate ≥93.0% (by GC)
Bis(2-ethylhexyl)maleate
Formule: C₂₀H₃₆O₄
Kookpunt: 173°C
Smeltpunt: -60°C
Vlampunt: 180°C
MDL Nummer: MFCD00027250
CAS nummer: 142-16-5
EINECS: 205-524-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3657abc9389b146f104c582d273c6f129f5478c7&vid=3657abc9389b146f104c582d273c6f129f5478c7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 93.0 %
Acid Value max. 0.1


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR