Methylmethacrylaat ≥99.8% (by GC) stabilised
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Methacrylic acid methyl ester
Stabilisatie: Stabilised with 6-tert-butyl-2,4-xylenol
M0087-25ML M0087-500ML
TCIAM0087-25MLEA 17.6 EUR
TCIAM0087-25ML TCIAM0087-500ML
Methylmethacrylaat ≥99.8% (by GC) stabilised
Methylmethacrylaat
Formule: CH₂=C(CH₃)COOCH₃
MW: 100.12 g/mol
Kookpunt: 100°C
Smeltpunt: -48
Vlampunt: 9°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00008587
CAS nummer: 80-62-6
EINECS: 201-297-1
UN: 1247
ADR: 3 II
Merck Index: 14,05941
Order Now
Specification Test Results
Appearance Colorless clear liquid
Purity (GC) min. 99.8 %