Dicoumarol ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Synoniemen: 3,3'-Methylenebis(4-hydroxycoumarin), Bishydroxycoumarin, Dicoumarin

M0216-1G M0216-25G
TCIAM0216-1GEA 25.3 EUR
TCIAM0216-1G TCIAM0216-25G
Dicoumarol ≥98.0% (by HPLC, titration analysis)
Dicoumarol
Formule: C₁₉H₁₂O₆
MW: 336.30 g/mol
MDL Nummer: MFCD00006857
CAS nummer: 66-76-2
EINECS: 200-632-9
UN: 2811
ADR: 6.1 III
Merck Index: 14,03090

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Solubility in 1mol/L NaOH almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR