Methylenebis(thioglycolic acid) ≥96.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

M0218-25G M0218-5G
TCIAM0218-25GEA 174 EUR
TCIAM0218-25G TCIAM0218-5G
Methylenebis(thioglycolic acid) ≥96.0% (by titrimetric analysis)
Methylenebis(thioglycolic acid)
Formule: C₅H₈O₄S₂
MDL Nummer: MFCD00135949
CAS nummer: 2068-24-8
EINECS: 218-189-5

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (Neutralization titration) min. 96.0 %
Melting point 125.0 to 129.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR