Methacrylic acid-tert-butyl ester monomer ≥98.0% (by GC) stabilised

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: Methacrylic acid tert-Butyl ester monomer, tert-Butyl Methacrylate

Stabilisatie: Stabilised with 4-methoxyphenol (MEHQ)

M0326-25ML M0326-500ML
TCIAM0326-25MLEA 18.7 EUR
TCIAM0326-25ML TCIAM0326-500ML
Methacrylic acid-tert-butyl ester monomer ≥98.0% (by GC) stabilised
Methacrylic acid-tert-butyl ester
Formule: C₈H₁₄O₂
MW: 142.1956 g/mol
Kookpunt: 136°C
Vlampunt: 30°C
MDL Nummer: MFCD00048245
CAS nummer: 585-07-9
EINECS: 209-548-7
UN: 3272
ADR: 3 III

Order Now

Specification Test Results

Appearance Colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR