Methylhydrazine sulphate ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

M0341-25G M0341-5G
TCIAM0341-25GEA 101 EUR
TCIAM0341-25G TCIAM0341-5G
Methylhydrazine sulphate ≥98.0% (by titrimetric analysis)
Methylhydrazine sulphate
Formule: CH₆N₂
MDL Nummer: MFCD00035423
CAS nummer: 302-15-8
EINECS: 206-115-4
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=75e75a16c559c3808135fcf0128d54b5308c2ed2&vid=75e75a16c559c3808135fcf0128d54b5308c2ed2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Potassium iodate Method) min. 98.0 %
Melting point 141.0 to 144.0 deg-C
Solubility in Water very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR