5-Methyl-2-pyrrolidone ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 5-Methylpyrrolidin-2-one, 5-Methyl-2-pyrrolidinone

M0406-10G M0406-25G
TCIAM0406-10GEA 74.8 EUR
TCIAM0406-10G TCIAM0406-25G
5-Methyl-2-pyrrolidone ≥98.0% (by GC)
5-Methyl-2-pyrrolidone
Formule: C₅H₉NO
Kookpunt: 142°C
MDL Nummer: MFCD00005273
CAS nummer: 108-27-0
EINECS: 203-566-9

Order Now

Specification Test Results

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR