4-Dimethylaminoazobenzene-4'-carboxylic acid ≥97.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: 4-Dimethylaminoazobenzene-4-carboxylic Acid, DABCYL acid, Para Methyl Red, p-Methyl Red

M0423-25G M0423-5G
TCIAM0423-25GEA 240 EUR
TCIAM0423-25G TCIAM0423-5G
4-Dimethylaminoazobenzene-4'-carboxylic acid ≥97.0% (by HPLC, titration analysis)
4-Dimethylaminoazobenzene-4'-carboxylic acid
Formule: C₁₅H₁₅N₃O₂
MDL Nummer: MFCD00058988
CAS nummer: 6268-49-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=263ab09b5e112dd27861e9e601185617c5a41d10&vid=263ab09b5e112dd27861e9e601185617c5a41d10&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Yellow to Amber to Dark red powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 97.0 %
Solubility in hot NaOH almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR